siRNA Details
Gene Symbol: Cers1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138647.3 | NP_619588.1 | 7 | 1131 | ||
GenBank mRNA | AK053885.1 | - | 7 | 1136 | ||
BC132318.1 | - | 7 | 1076 | 40130595 |
Gene Symbol: Gdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163282.2 | NP_001156754.1 | 7 | 1131 | ||
GenBank mRNA | AK162823.1 | - | 7 | 1114 | ||
BC079555.1 | - | 2 | 60 | 30359238 | ||
BC094351.1 | - | 7 | 1083 | |||
M62301.1 | - | 7 | 1119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
