siRNA Details
Gene Symbol: SMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000543.4 | NP_000534.3 | 3, 4 | 1448 | ||
NM_001007593.2 | NP_001007594.2 | 3, 4 | 1445 | |||
NM_001318087.1 | NP_001305016.1 | 3, 4 | 1448 | |||
NM_001318088.1 | NP_001305017.1 | 3, 4 | 1488 | |||
NR_134502.1 | 3, 4 | 795 | ||||
GenBank mRNA | AB209775.1 | - | 3, 4 | 1431 | ||
AK292388.1 | - | 3, 4 | 1379 | |||
AY649987.1 | - | 3, 4 | 627 | |||
HQ132745.1 | - | 3, 4 | 1257 | |||
HQ132746.1 | - | 3, 4 | 1188 | |||
HQ132747.1 | - | 3, 4 | 1257 | |||
HQ132748.1 | - | 3, 4 | 1291 | |||
M59916.1 | - | 3, 4 | 1357 | |||
X52678.1 | - | 2, 3 | 876 | |||
X59960.1 | - | 3, 4 | 1373 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
