siRNA Details
Gene Symbol: Bcas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166642.1 | NP_001160114.1 | 5 | 340 | ||
NM_001166643.1 | NP_001160115.1 | 4 | 254 | |||
NM_138681.4 | NP_619622.3 | 5 | 340 | |||
GenBank mRNA | AF313800.1 | - | 4 | 234 | ||
AK032423.1 | - | 5 | 291 | |||
AK034117.1 | - | 5 | 320 | |||
AK157222.1 | - | 5 | 291 | |||
AK160127.1 | - | 4 | 233 | |||
AK171964.1 | - | 4 | 207 | |||
BC028339.1 | - | 4 | 228 | |||
BC038173.1 | - | 3 | 197 | |||
BC050233.1 | - | 4 | 233 | |||
BC060690.1 | - | 4 | 217 | |||
BC080775.1 | - | 3 | 201 | |||
BC086693.1 | - | 4 | 221 | |||
BC118965.2 | - | 4 | 254 | 40097104 | ||
BC118966.1 | - | 4 | 236 | 40097105 | ||
BY116917.1 | - | 4 | 255 | |||
CN711546.1 | - | 5 | 323 |
