siRNA Details
Gene Symbol: Ppp2r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138748.5 | NP_620087.3 | 2 | 344 | ||
GenBank mRNA | AK046288.1 | - | 2 | 148 | ||
AK049184.1 | - | 2 | 335 | |||
AK150029.1 | - | 2 | 339 | |||
AK160980.1 | - | 2 | 199 | |||
AK179985.1 | - | 1 | 67 | |||
AK185428.1 | - | 1 | 67 | |||
AK207653.1 | - | 1 | 67 | |||
AK212725.1 | - | 1 | 67 | |||
AK215573.1 | - | 1 | 67 | |||
AK219416.1 | - | 1 | 67 | |||
AY035997.1 | - | 2 | 197 | |||
BC006626.1 | - | 2 | 260 | 3158086 | ||
BC052852.1 | - | 2 | 270 | 30058414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
