siRNA Details
Gene Symbol: Plxnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159521.2 | NP_001152993.1 | 27 | 4488 | ||
NM_001284506.1 | NP_001271435.1 | 26 | 4402 | |||
NM_138749.3 | NP_620088.2 | 27 | 4452 | |||
GenBank mRNA | AB093229.1 | - | 23 | 3026 | ||
AK147538.1 | - | 27 | 4453 | |||
AK154627.1 | - | 27 | 4403 | |||
BC003293.1 | - | 6 | 873 | |||
BC007481.1 | - | 24 | 3250 | |||
BC036274.1 | - | 15 | 2059 | |||
BC157960.1 | - | 27 | 4423 | 9088132 | ||
BC157961.1 | - | 27 | 4423 | 9088133 | ||
BC158086.1 | - | 27 | 4389 | 9088174 | ||
BC172162.1 | - | 27 | 4389 | 9054832 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
