siRNA Details
Gene Symbol: Ttc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138951.1 | NP_620401.1 | 1, 2 | 236 | ||
GenBank mRNA | AK149384.1 | - | 1, 2 | 267 | ||
AK165811.1 | - | 1, 2 | 134 | |||
BC021608.1 | - | 1, 2 | 148 | 5099426 | ||
BC021613.1 | - | 1, 2 | 242 | 5102393 | ||
BC024432.1 | - | 1, 2 | 236 | 4972715 | ||
BC026825.1 | - | 1, 2 | 183 | 4240199 |

More Information
Other Species Targeted

Gene Ontology Categories:
