siRNA Details
Gene Symbol: Ell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138953.2 | NP_620403.2 | 8 | 1633 | ||
GenBank mRNA | AK033541.1 | - | 3 | 621 | ||
AK140906.1 | - | 8 | 1587 | |||
AK144197.1 | - | 8 | 1598 | |||
AK144978.1 | - | 8 | 1597 | |||
AK145868.1 | - | 8 | 1582 | |||
AK154545.1 | - | 8 | 1594 | |||
AK156164.1 | - | 8 | 1600 | |||
BC006925.1 | - | 3 | 665 | |||
BC151188.1 | - | 8 | 1588 | 9088077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
