siRNA Details
Gene Symbol: Mthfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138745.2 | NP_620084.2 | 7, 8 | 895 | ||
GenBank mRNA | AF364579.1 | - | 7, 8 | 676 | ||
AK042019.1 | - | 7, 8 | 775 | |||
AK042958.1 | - | 7, 8 | 731 | |||
AK088700.1 | - | 7, 8 | 692 | |||
AK129012.1 | - | 7, 8 | 829 | |||
AK133848.1 | - | 6, 7 | 630 | |||
AK159812.1 | - | 7, 8 | 707 | |||
AK165951.1 | - | 7, 8 | 727 | |||
AK208410.1 | - | 1, 2 | 121 | |||
BC008523.1 | - | 7, 8 | 719 | 3590449 | ||
BC013563.1 | - | 1, 2 | 32 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
