siRNA Details
Gene Symbol: Plxnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159521.2 | NP_001152993.1 | 38 | 6233 | ||
NM_001284506.1 | NP_001271435.1 | 37 | 6147 | |||
NM_138749.3 | NP_620088.2 | 38 | 6197 | |||
GenBank mRNA | AB093229.1 | - | 34 | 4812 | ||
AK003544.1 | - | 1 | 234 | |||
AK147538.1 | - | 38 | 6198 | |||
AK154627.1 | - | 38 | 6148 | |||
BC003293.1 | - | 17 | 2618 | |||
BC007481.1 | - | 35 | 4995 | |||
BC036274.1 | - | 26 | 3804 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
