siRNA Details
Gene Symbol: Ell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138953.2 | NP_620403.2 | 10 | 1872 | ||
GenBank mRNA | AK140906.1 | - | 10 | 1826 | ||
AK144197.1 | - | 10 | 1837 | |||
AK144978.1 | - | 10 | 1836 | |||
AK145868.1 | - | 10 | 1821 | |||
AK154545.1 | - | 10 | 1833 | |||
AK156164.1 | - | 10 | 1839 | |||
AK161961.1 | - | 3 | 322 | |||
AK163470.1 | - | 1 | 1863 | |||
BC006925.1 | - | 5 | 904 | |||
BC151188.1 | - | 10 | 1827 | 9088077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
