siRNA Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF402611.1 | - | 3, 4 | 418 | ||
AF402612.1 | - | 2, 3 | 315 | |||
AF402613.1 | - | 3, 4 | 465 | |||
AK030390.1 | - | 3, 4 | 568 | |||
AK043656.1 | - | 3, 4 | 463 | |||
AK054242.1 | - | 3, 4 | 406 | |||
AK134961.1 | - | 3, 4 | 481 | |||
AK149626.1 | - | 3, 4 | 469 | |||
BB617683.1 | - | 3, 4 | 568 | |||
BC028644.1 | - | 3, 4 | 340 | |||
BC129902.1 | - | 3, 4 | 505 | 40111637 | ||
BC129903.1 | - | 3, 4 | 506 | 40111639 | ||
BC145523.1 | - | 3, 4 | 417 |
