siRNA Details
Gene Symbol: HGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000601.5 | NP_000592.3 | 18 | 2628 | ||
NM_001010932.2 | NP_001010932.1 | 18 | 2613 | |||
XM_006715956.2 | XP_006716019.1 | 19 | 2591 | |||
XM_011516115.2 | XP_011514417.1 | 19 | 2576 | |||
GenBank mRNA | AK291674.1 | - | 18 | 2563 | ||
M29145.1 | - | 18 | 2502 | |||
M73239.1 | - | 18 | 2466 | |||
M73240.1 | - | 18 | 2451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
