siRNA Details
Gene Symbol: Zfp622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144523.2 | NP_653106.1 | 1 | 808 | ||
GenBank mRNA | AK031232.1 | - | 1 | 808 | ||
AK149106.1 | - | 1 | 739 | |||
AK154951.1 | - | 1 | 734 | |||
AK170489.1 | - | 1 | 807 | |||
BC006964.1 | - | 1 | 713 | 3153893 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
