siRNA Details
Gene Symbol: CXCL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000609.6 | NP_000600.1 | 4 | 2599 | ||
NM_001277990.1 | NP_001264919.1 | 3 | 2189 | |||
GenBank mRNA | AK124641.1 | - | 1 | 3670 | ||
AL713778.1 | - | 1 | 3459 | |||
BX647204.1 | - | 1 | 4400 | |||
L36033.1 | - | 4 | 2586 | |||
U16752.1 | - | 4 | 2585 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
