siRNA Details
Gene Symbol: Bre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144541.1 | NP_653124.1 | 2 | 138 | ||
GenBank mRNA | AF440752.1 | - | 2 | 138 | ||
AK020047.1 | - | 2 | 224 | |||
AK050695.1 | - | 2 | 183 | |||
AK080991.1 | - | 2 | 174 | |||
AK156929.1 | - | 2 | 230 | |||
AK166884.1 | - | 2 | 174 | |||
AK177683.1 | - | 2 | 137 | |||
AK207416.1 | - | 2 | 137 | |||
AK207670.1 | - | 2 | 137 | |||
AK216664.1 | - | 1 | 144 | |||
BC029277.1 | - | 2 | 489 | |||
BC061000.1 | - | 2 | 214 | 30305062 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
