siRNA Details
Gene Symbol: Esco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081222.1 | NP_001074691.1 | 3 | 2468 | ||
GenBank mRNA | AK139549.1 | - | 3 | 2192 | ||
AK145333.1 | - | 3 | 2469 | |||
AK173297.1 | - | 1 | 1848 | |||
BC008220.1 | - | 1 | 218 | |||
BC076613.1 | - | 3 | 2198 | |||
BC151069.1 | - | 3 | 2424 | 9087982 | ||
BC151077.1 | - | 3 | 2424 | 9087990 | ||
BC172153.1 | - | 3 | 2424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
