siRNA Details
Gene Symbol: Prdm15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144789.2 | NP_659038.2 | 20 | 2662 | ||
GenBank mRNA | AK144820.1 | - | 20 | 2567 | ||
AY063457.1 | - | 1 | 220 | |||
BC046433.1 | - | 1 | 221 | |||
BC072597.1 | - | 18 | 2349 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
