siRNA Details
Gene Symbol: Trim24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272064.1 | NP_001258993.1 | 2, 3 | 791 | ||
NM_001272076.1 | NP_001259005.1 | 2, 3 | 371 | |||
NM_145076.4 | NP_659542.3 | 2, 3 | 791 | |||
GenBank mRNA | AB221637.1 | - | 2, 3 | 491 | ||
AK084870.1 | - | 2, 3 | 783 | |||
AK133144.1 | - | 2, 3 | 371 | |||
BC056959.1 | - | 2, 3 | 769 | 5698079 | ||
S78219.1 | - | 2, 3 | 791 | |||
S78221.1 | - | 2, 3 | 791 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
