siRNA Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF402611.1 | - | 4, 5 | 933 | ||
AF402612.1 | - | 3, 4 | 830 | |||
AF402613.1 | - | 4, 5 | 980 | |||
AK034504.1 | - | 3, 4 | 923 | |||
AK043656.1 | - | 4, 5 | 978 | |||
AK054242.1 | - | 4, 5 | 921 | |||
AK134961.1 | - | 4, 5 | 996 | |||
AK149626.1 | - | 4, 5 | 984 | |||
BC129902.1 | - | 4, 5 | 1020 | 40111637 | ||
BC129903.1 | - | 4, 5 | 1021 | 40111639 | ||
BC145523.1 | - | 4, 5 | 1074 |
