siRNA Details
Gene Symbol: Trim24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272064.1 | NP_001258993.1 | 5 | 1170 | ||
NM_001272076.1 | NP_001259005.1 | 5 | 750 | |||
NM_145076.4 | NP_659542.3 | 5 | 1170 | |||
GenBank mRNA | AB221637.1 | - | 5 | 870 | ||
AK080890.1 | - | 4 | 1023 | |||
AK084870.1 | - | 5 | 1160 | |||
AK133144.1 | - | 5 | 750 | |||
BC056959.1 | - | 5 | 1148 | 5698079 | ||
S78219.1 | - | 5 | 1170 | |||
S78221.1 | - | 5 | 1170 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
