siRNA Details
Gene Symbol: SOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000636.3 | NP_000627.2 | 2, 3 | 394 | ||
NM_001024465.2 | NP_001019636.1 | 2, 3 | 394 | |||
NM_001322815.1 | NP_001309744.1 | 2, 3 | 394 | |||
NM_001322817.1 | NP_001309746.1 | 4, 5 | 1923 | |||
NM_001322819.1 | NP_001309748.1 | 2, 3 | 300 | |||
NM_001322820.1 | NP_001309749.1 | 2, 3 | 303 | |||
GenBank mRNA | AK097395.1 | - | 4, 5 | 1920 | ||
AK304766.1 | - | 2, 3 | 307 | |||
AK313082.1 | - | 2, 3 | 259 | |||
AY280718.1 | - | 2, 3 | 230 | |||
AY280719.1 | - | 1, 2 | 222 | |||
AY280720.1 | - | 1, 2 | 221 | |||
AY280721.1 | - | 1, 2 | 221 | |||
BC012423.1 | - | 2, 3 | 290 | 3880111 | ||
BC035422.1 | - | 2, 3 | 290 | |||
BG699596.1 | - | 2, 3 | 394 | |||
BP289103.1 | - | 2, 3 | 301 | |||
BT006967.1 | - | 2, 3 | 233 | |||
BU164685.1 | - | 2, 3 | 394 | |||
DA094180.1 | - | 2, 3 | 304 | |||
DA318772.1 | - | 2, 3 | 390 | |||
DQ003134.1 | - | 1, 2 | 230 | |||
M36693.1 | - | 2, 3 | 327 | |||
X07834.1 | - | 2, 3 | 237 | |||
X14322.1 | - | 2, 3 | 328 | |||
X15132.1 | - | 2, 3 | 293 | |||
X59445.1 | - | 2, 3 | 293 | |||
Y00472.1 | - | 2, 3 | 299 | |||
Y00985.1 | - | 2, 3 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
