siRNA Details
Gene Symbol: Esco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081222.1 | NP_001074691.1 | 8, 9 | 3081 | ||
GenBank mRNA | AK040576.1 | - | 8, 9 | 909 | ||
AK049589.1 | - | 7, 8 | 797 | |||
AK139549.1 | - | 8, 9 | 2805 | |||
AK145333.1 | - | 8, 9 | 3082 | |||
AK145870.1 | - | 7, 8 | 729 | |||
BC008220.1 | - | 7, 8 | 953 | |||
BC076613.1 | - | 8, 9 | 2811 | |||
BC151069.1 | - | 8, 9 | 3037 | 9087982 | ||
BC151077.1 | - | 8, 9 | 3037 | 9087990 | ||
BC172153.1 | - | 9, 10 | 3159 |

More Information
Other Species Targeted

Gene Ontology Categories:
