siRNA Details
Gene Symbol: Eif3j1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144545.4 | NP_653128.2 | 8 | 1358 | ||
GenBank mRNA | AK019016.1 | - | 1 | 114 | ||
AK082048.1 | - | 7 | 1149 | |||
AK137221.1 | - | 1 | 136 | |||
AK147154.1 | - | 9 | 1204 | |||
AK148004.1 | - | 8 | 1361 | |||
AK154934.1 | - | 8 | 1359 | |||
AK161038.1 | - | 8 | 1316 | |||
BC006055.1 | - | 4 | 935 |
Gene Symbol: Eif3j2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256055.1 | NP_001242984.1 | 1 | 1365 | ||
GenBank mRNA | BC080724.1 | - | 1 | 868 | ||
BC080788.1 | - | 1 | 1319 | 4166214 | ||
BC108355.1 | - | 1 | 1145 |
