siRNA Details
Gene Symbol: Prdm15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144789.2 | NP_659038.2 | 23 | 2991 | ||
GenBank mRNA | AK053449.1 | - | 1 | 46 | ||
AK144820.1 | - | 23 | 2896 | |||
AY063457.1 | - | 4 | 549 | |||
BC046433.1 | - | 4 | 550 | |||
BC072597.1 | - | 21 | 2678 |

More Information
Other Species Targeted

Gene Ontology Categories:
