siRNA Details
Gene Symbol: CDK14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287135.1 | NP_001274064.1 | 3 | 475 | ||
NM_001287136.1 | NP_001274065.1 | 2 | 228 | |||
NM_001287137.1 | NP_001274066.1 | 2 | 228 | |||
NM_012395.3 | NP_036527.1 | 2 | 299 | |||
XM_005250436.3 | XP_005250493.1 | 4 | 274 | |||
XM_005250438.3 | XP_005250495.1 | 2 | 221 | |||
XM_005250439.2 | XP_005250496.1 | 3 | 1032 | |||
XM_011516306.2 | XP_011514608.1 | 3 | 843 | |||
XM_011516307.2 | XP_011514609.1 | 3 | 843 | |||
XM_017012320.1 | XP_016867809.1 | 4 | 407 | |||
XM_017012321.1 | XP_016867810.1 | 2 | 136 | |||
XM_017012322.1 | XP_016867811.1 | 2 | 778 | |||
GenBank mRNA | AB020641.1 | - | 2 | 299 | ||
AF119833.1 | - | 3 | 347 | |||
AK289782.1 | - | 2 | 157 | |||
AK295086.1 | - | 2 | 211 | |||
AK296406.1 | - | 2 | 182 | |||
AK297974.1 | - | 3 | 431 | |||
AK307484.1 | - | 2 | 182 | |||
AK316026.1 | - | 2 | 182 | |||
BC136476.1 | - | 2 | 369 | 9020465 | ||
BC136477.1 | - | 2 | 254 | 9020466 | ||
BC152388.1 | - | 2 | 158 | |||
BC152436.1 | - | 2 | 299 | 8862558 | ||
BC167152.1 | - | 2 | 299 | 8862547 | ||
BC167156.1 | - | 2 | 299 | 8862570 | ||
DC325068.1 | - | 2 | 231 | |||
DC420282.1 | - | 3 | 466 | |||
DQ655962.2 | - | 4 | 255 | |||
DQ655963.2 | - | 3 | 209 | |||
DQ655965.2 | - | 4 | 281 | |||
DQ655966.2 | - | 5 | 310 | |||
DQ656060.2 | - | 6 | 448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
