siRNA Details
Gene Symbol: Tnrc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144812.2 | NP_659061.2 | 25 | 5390 | ||
NM_177124.4 | NP_796098.3 | 22 | 5181 | |||
GenBank mRNA | AK051174.1 | - | 7 | 778 | ||
AK052247.1 | - | 17 | 2144 | |||
AK122440.1 | - | 11 | 1382 | |||
AK162702.1 | - | 16 | 1881 | |||
AK162788.1 | - | 17 | 2440 | |||
BC017531.1 | - | 4 | 884 | |||
BC080750.1 | - | 6 | 605 | |||
BC117548.1 | - | 4 | 642 | |||
BC118054.1 | - | 4 | 642 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
