siRNA Details
Gene Symbol: ACACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198834.2 | NP_942131.1 | 3 | 721 | ||
NM_198836.2 | NP_942133.1 | 3 | 289 | |||
NM_198839.2 | NP_942136.1 | 7 | 1395 | |||
XM_005257267.4 | XP_005257324.1 | 3 | 202 | |||
XM_006721853.1 | XP_006721916.1 | 3 | 732 | |||
XM_011524701.1 | XP_011523003.1 | 3 | 642 | |||
XM_011524703.1 | XP_011523005.1 | 3 | 274 | |||
XM_011524704.2 | XP_011523006.1 | 3 | 207 | |||
XM_017024553.1 | XP_016880042.1 | 4 | 756 | |||
XM_017024554.1 | XP_016880043.1 | 4 | 756 | |||
XM_017024555.1 | XP_016880044.1 | 3 | 355 | |||
GenBank mRNA | AB371587.1 | - | 3 | 268 | ||
AJ564444.1 | - | 3 | 297 | |||
AK308905.1 | - | 3 | 583 | |||
AK309084.1 | - | 3 | 277 | |||
AY237919.1 | - | 1 | 98 | |||
AY315621.1 | - | 3 | 289 | |||
AY315622.1 | - | 3 | 289 | |||
AY315623.1 | - | 7 | 1365 | |||
AY315625.1 | - | 3 | 691 | |||
AY315626.1 | - | 3 | 691 | |||
AY315627.1 | - | 1 | 98 | |||
BC137287.1 | - | 3 | 226 | 9021284 | ||
U19822.1 | - | 1 | 98 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
