siRNA Details
Gene Symbol: Ice1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144837.3 | NP_659086.2 | 15 | 6118 | ||
GenBank mRNA | AK129244.1 | - | 3 | 4555 | ||
AK134564.1 | - | 3 | 2561 | |||
BC018507.1 | - | 2 | 213 | |||
BC026613.1 | - | 3 | 644 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
