siRNA Details
Gene Symbol: Ugt3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144845.3 | NP_659094.1 | 2 | 250 | ||
GenBank mRNA | AK041045.1 | - | 2 | 235 | ||
AK050128.1 | - | 2 | 251 | |||
AK052644.1 | - | 2 | 260 | |||
AK143815.1 | - | 2 | 265 | |||
BC022134.1 | - | 2 | 235 | 5098754 | ||
BC034837.1 | - | 2 | 231 | 5134131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
