siRNA Details
Gene Symbol: Msto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144898.2 | NP_659147.2 | 2 | 262 | ||
GenBank mRNA | AK139708.1 | - | 245 | |||
AK163280.1 | - | 2 | 122 | |||
AK166236.1 | - | 2 | 234 | |||
AK166834.1 | - | 1 | 306 | |||
AK167586.1 | - | 2 | 210 | |||
AK170160.1 | - | 2 | 245 | |||
BC008103.1 | - | 2 | 212 | 3966815 | ||
BC108354.1 | - | 2 | 196 | 5035560 | ||
BY267786.1 | - | 2 | 263 |
