siRNA Details
Gene Symbol: Tnrc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144812.2 | NP_659061.2 | 25 | 5712 | ||
NM_177124.4 | NP_796098.3 | 22 | 5503 | |||
GenBank mRNA | AK051174.1 | - | 7 | 1100 | ||
AK052247.1 | - | 17 | 2466 | |||
AK122440.1 | - | 11 | 1704 | |||
AK162702.1 | - | 16 | 2203 | |||
AK162788.1 | - | 17 | 2762 | |||
BC017531.1 | - | 4 | 1206 | |||
BC080750.1 | - | 6 | 927 | |||
BC117548.1 | - | 4 | 964 | |||
BC118054.1 | - | 4 | 964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
