siRNA Details
Gene Symbol: Ice1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144837.3 | NP_659086.2 | 16 | 6249 | ||
GenBank mRNA | AK129244.1 | - | 4 | 4686 | ||
AK134564.1 | - | 4 | 2692 | |||
BC018507.1 | - | 3 | 344 | |||
BC026613.1 | - | 4 | 775 |

More Information
Other Species Targeted

Gene Ontology Categories:
