siRNA Details
Gene Symbol: Dnah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033668.1 | NP_001028840.1 | 2 | 281 | ||
GenBank mRNA | AK053204.1 | - | 2 | 277 | ||
BC023155.1 | - | 2 | 307 |

More Information
Other Species Targeted

Gene Ontology Categories:
