siRNA Details
Gene Symbol: Zdhhc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144887.4 | NP_659136.1 | 4 | 1472 | ||
GenBank mRNA | AK028737.1 | - | 1 | 38 | ||
AK082513.1 | - | 4 | 1397 | |||
AK145086.1 | - | 4 | 1399 | |||
AK173251.1 | - | 4 | 1427 | |||
AY894891.1 | - | 4 | 1464 | |||
BC020051.1 | - | 3 | 442 | 3987696 | ||
BC065155.1 | - | 4 | 1342 | 5706087 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
