siRNA Details
Gene Symbol: Psmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032644.1 | - | 2 | 351 | ||
AK045891.1 | - | 2 | 363 | |||
AK053924.1 | - | 2 | 381 | |||
AK078867.1 | - | 2 | 339 | |||
BC012260.1 | - | 2 | 342 | |||
BC128314.1 | - | 1 | 125 | |||
BC131928.1 | - | 2 | 244 | 40130205 | ||
BC131930.1 | - | 2 | 244 | 40130207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
