siRNA Details
Gene Symbol: CDK14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287135.1 | NP_001274064.1 | 15 | 5001 | ||
NM_001287136.1 | NP_001274065.1 | 14 | 4754 | |||
NM_001287137.1 | NP_001274066.1 | 13 | 4659 | |||
NM_012395.3 | NP_036527.1 | 14 | 4825 | |||
XM_005250436.3 | XP_005250493.1 | 16 | 4800 | |||
XM_005250438.3 | XP_005250495.1 | 14 | 4747 | |||
XM_005250439.2 | XP_005250496.1 | 15 | 5558 | |||
XM_017012320.1 | XP_016867809.1 | 16 | 4933 | |||
XM_017012321.1 | XP_016867810.1 | 14 | 4662 | |||
GenBank mRNA | AB020641.1 | - | 14 | 4829 | ||
BC152436.1 | - | 14 | 4829 | 8862558 | ||
BC167152.1 | - | 14 | 4829 | 8862547 | ||
BC167156.1 | - | 14 | 4829 | 8862570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
