siRNA Details
Gene Symbol: Tnrc6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144812.2 | NP_659061.2 | 25 | 5838 | ||
NM_177124.4 | NP_796098.3 | 22 | 5629 | |||
GenBank mRNA | AK051174.1 | - | 7 | 1226 | ||
AK052247.1 | - | 17 | 2592 | |||
AK122440.1 | - | 11 | 1829 | |||
AK162702.1 | - | 16 | 2329 | |||
AK162788.1 | - | 17 | 2889 | |||
BC017531.1 | - | 4 | 1332 | |||
BC080750.1 | - | 6 | 1053 | |||
BC117548.1 | - | 4 | 1090 | |||
BC118054.1 | - | 4 | 1091 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_138845, XM_006242063, XM_006242061, XM_006242064, BC061751.
Gene Ontology Categories:
