siRNA Details
Gene Symbol: Cyyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144853.3 | NP_659102.1 | 4 | 855 | ||
GenBank mRNA | AK031840.1 | - | 4 | 879 | ||
AY061854.1 | - | 4 | 877 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
