siRNA Details
Gene Symbol: BC021614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144869.2 | NP_659118.1 | 2 | 139 | ||
GenBank mRNA | BC021614.1 | - | 2 | 139 | 5102505 | |
BC034269.1 | - | 3 | 152 | 4196879 |

More Information
Other Species Targeted

Gene Ontology Categories:
