siRNA Details
Gene Symbol: Ppp2r5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144880.4 | NP_659129.2 | 13 | 2960 | ||
GenBank mRNA | AK145397.1 | - | 14 | 2346 | ||
BC023062.1 | - | 7 | 1529 | |||
BC034058.1 | - | 3 | 1376 | |||
BC059026.1 | - | 13 | 2958 | 6810754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
