siRNA Details
Gene Symbol: CDK14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287135.1 | NP_001274064.1 | 3 | 433 | ||
NM_001287136.1 | NP_001274065.1 | 2 | 186 | |||
NM_001287137.1 | NP_001274066.1 | 2 | 186 | |||
NM_012395.3 | NP_036527.1 | 2 | 257 | |||
XM_005250436.3 | XP_005250493.1 | 4 | 232 | |||
XM_005250438.3 | XP_005250495.1 | 2 | 179 | |||
XM_005250439.2 | XP_005250496.1 | 3 | 990 | |||
XM_011516306.2 | XP_011514608.1 | 3 | 801 | |||
XM_011516307.2 | XP_011514609.1 | 3 | 801 | |||
XM_017012320.1 | XP_016867809.1 | 4 | 365 | |||
XM_017012321.1 | XP_016867810.1 | 2 | 94 | |||
XM_017012322.1 | XP_016867811.1 | 2 | 736 | |||
GenBank mRNA | AB020641.1 | - | 2 | 257 | ||
AF119833.1 | - | 3 | 305 | |||
AK289782.1 | - | 2 | 115 | |||
AK295086.1 | - | 2 | 169 | |||
AK296406.1 | - | 2 | 140 | |||
AK297974.1 | - | 3 | 389 | |||
AK307484.1 | - | 2 | 140 | |||
AK316026.1 | - | 2 | 140 | |||
BC136476.1 | - | 2 | 327 | 9020465 | ||
BC136477.1 | - | 2 | 212 | 9020466 | ||
BC152388.1 | - | 2 | 116 | |||
BC152436.1 | - | 2 | 257 | 8862558 | ||
BC167152.1 | - | 2 | 257 | 8862547 | ||
BC167156.1 | - | 2 | 257 | 8862570 | ||
DC325068.1 | - | 2 | 189 | |||
DC420282.1 | - | 3 | 424 | |||
DQ655962.2 | - | 4 | 213 | |||
DQ655963.2 | - | 3 | 167 | |||
DQ655965.2 | - | 4 | 239 | |||
DQ655966.2 | - | 5 | 268 | |||
DQ656060.2 | - | 6 | 406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
