siRNA Details
Gene Symbol: Tmem184a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161548.1 | NP_001155020.1 | 8 | 1229 | ||
NM_144914.3 | NP_659163.3 | 7 | 1049 | |||
GenBank mRNA | AK050489.1 | - | 8 | 1199 | ||
AK148454.1 | - | 1021 | ||||
BC019731.1 | - | 3 | 438 | 3993397 | ||
BC026659.1 | - | 8 | 1188 | 4988857 |
