siRNA Details
Gene Symbol: Tnrc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144925.3 | NP_659174.3 | 18 | 5018 | ||
GenBank mRNA | AK044679.1 | - | 6 | 1402 | ||
AK138426.1 | - | 8 | 1068 | |||
AK142808.1 | - | 3 | 387 | |||
AK142809.1 | - | 3 | 387 | |||
AK147327.1 | - | 18 | 4723 | |||
BC005741.1 | - | 7 | 853 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107549, XM_008759749, XM_006230190, XM_008759747, XM_008759748.
Gene Ontology Categories:
