siRNA Details
Gene Symbol: Cd207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144943.3 | NP_659192.2 | 2 | 185 | ||
GenBank mRNA | AK142731.1 | - | 2 | 127 | ||
AY078415.1 | - | 2 | 127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
