siRNA Details
Gene Symbol: Ddx60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293783.1 | NP_001280712.1 | 30, 31 | 4454 | ||
GenBank mRNA | BC013672.1 | - | 3, 4 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
