siRNA Details
Gene Symbol: Eif4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159375.1 | NP_001152847.1 | 4 | 427 | ||
NM_144958.4 | NP_659207.1 | 4 | 427 | |||
GenBank mRNA | AK010644.1 | - | 4 | 329 | ||
AK011529.1 | - | 4 | 320 | |||
AK077429.1 | - | 4 | 320 | |||
AK088124.1 | - | 3 | 664 | |||
AK135752.1 | - | 4 | 320 | |||
AK135994.1 | - | 4 | 321 | |||
AK144850.1 | - | 4 | 320 | |||
AK145375.1 | - | 4 | 322 | |||
AK145508.1 | - | 4 | 322 | |||
AK146424.1 | - | 4 | 322 | |||
AK146464.1 | - | 4 | 320 | |||
AK152210.1 | - | 1 | 103 | |||
AK152694.1 | - | 4 | 322 | |||
AK159643.1 | - | 4 | 322 | |||
AK162017.1 | - | 4 | 322 | |||
AK166439.1 | - | 4 | 320 | |||
AK166527.1 | - | 4 | 320 | |||
AK166572.1 | - | 4 | 322 | |||
AK166674.1 | - | 4 | 322 | |||
AK167003.1 | - | 4 | 320 | |||
AK167083.1 | - | 4 | 320 | |||
AK167196.1 | - | 4 | 322 | |||
AK167205.1 | - | 4 | 319 | |||
AK167906.1 | - | 4 | 322 | |||
AK167988.1 | - | 4 | 320 | |||
AK168780.1 | - | 4 | 320 | |||
AK169023.1 | - | 4 | 320 | |||
AK169030.1 | - | 4 | 313 | |||
AK169057.1 | - | 4 | 320 | |||
AK169156.1 | - | 4 | 320 | |||
AK169279.1 | - | 4 | 320 | |||
AK179026.1 | - | 3 | 250 | |||
AK182003.1 | - | 2 | 250 | |||
AK207714.1 | - | 3 | 250 | |||
AK210015.1 | - | 3 | 250 | |||
AK211426.1 | - | 251 | ||||
BC012282.1 | - | 4 | 309 | |||
BC049915.1 | - | 4 | 310 | 5068709 | ||
BC099392.1 | - | 3 | 303 | |||
X03039.1 | - | 4 | 317 | |||
X03040.1 | - | 3 | 320 |

More Information
Other Species Targeted

Gene Ontology Categories:
