siRNA Details
Gene Symbol: 2610305D13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011986.1 | - | 2 | 421 | ||
BC032186.1 | - | 1 | 207 | |||
BC058858.1 | - | 5 | 724 | |||
BC138179.1 | - | 2 | 362 | 8861198 | ||
BC145854.1 | - | 2 | 362 | 40131157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
