siRNA Details
Gene Symbol: DHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000791.3 | NP_000782.1 | 6 | 1639 | ||
NM_001290354.1 | NP_001277283.1 | 5 | 1589 | |||
NM_001290357.1 | NP_001277286.1 | 5 | 1523 | |||
NR_110936.1 | 4 | 1462 | ||||
GenBank mRNA | AK293146.1 | - | 5 | 1167 | ||
AK297302.1 | - | 5 | 1094 | |||
BC000192.1 | - | 6 | 1202 | 3352539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
