siRNA Details
Gene Symbol: DIO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000792.6 | NP_000783.2 | 1 | 336 | ||
NM_001039715.2 | NP_001034804.1 | 1 | 336 | |||
NM_001039716.2 | NP_001034805.1 | 1 | 336 | |||
NM_001324316.1 | NP_001311245.1 | 1 | 336 | |||
NR_136693.1 | 2 | 253 | ||||
GenBank mRNA | AK290780.1 | - | 1 | 336 | ||
AY560374.1 | - | 1 | 312 | |||
AY560377.1 | - | 1 | 312 | |||
AY560378.1 | - | 1 | 312 | |||
AY560380.1 | - | 2 | 188 | |||
AY560381.1 | - | 1 | 312 | |||
AY560382.1 | - | 2 | 229 | |||
AY560383.1 | - | 2 | 229 | |||
BC017955.1 | - | 1 | 335 | 4607072 | ||
BC107170.2 | - | 1 | 335 | 40036824 | ||
BC107171.2 | - | 1 | 335 | 40036826 | ||
FJ002243.1 | - | 2 | 251 | |||
FJ002248.1 | - | 1 | 334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
