siRNA Details
Gene Symbol: Daglb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144915.3 | NP_659164.2 | 15 | 2256 | ||
GenBank mRNA | AK081509.1 | - | 894 | |||
AK086747.1 | - | 15 | 2168 | |||
AK087884.1 | - | 15 | 2111 | |||
AK136372.1 | - | 15 | 2114 | |||
BC016105.1 | - | 15 | 2152 | 4921222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
